VWR® Wide-Mouth Flask
High Quality, Great Value.
- Borosilicate glass
- Uniform wall thickness
- White enamel metric scale
- Marking spot
Each flask can be heated and used with acids, salt solutions, and organic solvents.
Frequently Bought Together
High Quality, Great Value.
Each flask can be heated and used with acids, salt solutions, and organic solvents.