VWR® Volumetric Flasks
Supplier: VWR International
Jump to:
Acerca de este artículo
Our borosilicate flasks meet ASTM standards.
- Class A Flasks
- Borosilicate Glass Construction
- Graduated Marking for Measurements
- Economical
- Snap-Caps Included
The necks fit the included snap-caps.
The Class A, volumetric flasks include a graduation line for accurate measurements.
6 Options Available Below
Product Details & Documents
Our borosilicate flasks meet ASTM standards.
- Class A Flasks
- Borosilicate Glass Construction
- Graduated Marking for Measurements
- Economical
- Snap-Caps Included
The necks fit the included snap-caps.
The Class A, volumetric flasks include a graduation line for accurate measurements.
Certificates
Buscar por número de loteProduct Family Options
Información del producto
- CapacityAvailabilityPrice
- 50 mlEach (1UN)$3.30Case de 12 (1UN)$39.60
- 100 mlEach (1UN)$122.56Case de 12 (1UN)$133.59
- 250 mlEach (1UN)$183.27Case de 12 (1UN)$199.77
- 500 mlEach (1UN)$237.01Case de 12 (1UN)$258.34
- 1000 mlEach (1UN)$390.74Case de 6 (1UN)$425.91
- 2000 mlEach (1UN)$626.14Case de 4 (1UN)$682.49