VWR® Micropipette Tip Reload System
Acerca de este artículo
Save time, plastic, and money with the Micropipette tip reload system.
- Tips available in three different configuarations
- Save plastic and re-use VWR® hinged rack boxes with the Reload Sleeves
- Flip-lock boxes offer easy one-handed operation
- Lid lip enables stable stacking
- Tips work with most manufacturer's micropipettors
The reloading operation features sleeve alignment, which orients loading for a perfect fit every time.
The tongue and groove format keeps the insert fastened while applying tips, and the tall clip makes the insert easy to handle without touching tips with your hand.
Packaging: The Racked Tips contain 10 boxes ('racks') of 96 per pack. There are 5 packs per case.
The Reload Sleeves contain 10 inserts of 96 tips per pack. There are 5 packs per case.
The Bulk Bags contain 1000 loose tips per bag. There are 10 bags per case.
10 Options Available Below
Product Details & Documents
Save time, plastic, and money with the Micropipette tip reload system.
- Tips available in three different configuarations
- Save plastic and re-use VWR® hinged rack boxes with the Reload Sleeves
- Flip-lock boxes offer easy one-handed operation
- Lid lip enables stable stacking
- Tips work with most manufacturer's micropipettors
The reloading operation features sleeve alignment, which orients loading for a perfect fit every time.
The tongue and groove format keeps the insert fastened while applying tips, and the tall clip makes the insert easy to handle without touching tips with your hand.
Packaging: The Racked Tips contain 10 boxes ('racks') of 96 per pack. There are 5 packs per case.
The Reload Sleeves contain 10 inserts of 96 tips per pack. There are 5 packs per case.
The Bulk Bags contain 1000 loose tips per bag. There are 10 bags per case.
Documents
Certificates
Buscar por número de loteProduct Family Options
Racked Tips
Información del producto
- VolumeTip ColorPackedAvailabilityPrice
- Discontinued-
- 0.1 - 10 µlNatural10 Racks/Pack, 5 Packs/CasePaquete de 10$2,270.00Case de 5 (10UN)$1,853.53
- 1 - 200 µlNatural10 Racks/Pack, 5 Packs/CasePaquete de 10$1,880.00Case de 5 (10UN)$1,536.59
- 1 - 200 µlNatural10 Racks/Pack, 5 Packs/CasePaquete de 10$1,880.00Case de 5 (10UN)$1,536.59
Bulk Bags
Información del producto
- VolumeTip ColorPackedAvailabilityPrice
- 1 - 200 µlNatural1000 Tips/Bag, 10 Bags/CasePaquete de 1,000$777.00Case de 10 (1,000,UN)$1,060.36Supplier Number: 113-GN
- 100 - 1250 µlNatural1000 Tips/Bag, 5 Bags/CasePaquete de 1,000$921.00Case de 10 (1,000,UN)$1,256.58Supplier Number: 112NXL
- 0.1 - 10 µlNatural1000 Tips/Bag, 10 Bags/CasePaquete de 1,000$573.00Case de 10 (1,000,UN)$782.08Supplier Number: 104
- 0.1 - 10 µlNatural1000 Tips/Bag, 10 Bags/CasePaquete de 1,000$573.00Case de 10 (1,000,UN)$782.08Supplier Number: 104
- 1 - 200 µlNatural1000 Tips/Bag, 10 Bags/CasePaquete de 1,000$777.00Case de 10 (1,000,UN)$1,060.36Supplier Number: 113-GN
- 100 - 1250 µlNatural1000 Tips/Bag, 10 Bags/CasePaquete de 1,000$921.00Case de 10 (1,000,UN)$1,256.58Supplier Number: 112NXL
Reload Sleeves
Información del producto
- VolumeTip ColorPackedAvailabilityPrice
- 0.1 - 10 µlNatural10 Inserts/Pack, 5 Packs/CaseDiscontinued-Supplier Number: T104RL-V
- 1 - 200 µlNatural10 Inserts/Pack, 5 Packs/CaseDiscontinued-Supplier Number: T113RLN-V
- 100 - 1250 µlNatural10 Inserts/Pack, 5 Packs/CasePaquete de 10$1,860.00Case de 5 (10UN)$1,518.46Supplier Number: T112NXLRL-V