VWR® SoftCIDE® Extra-Mild Hand Soap
VWR® softcide® extra-mild hand soap
Catalog # 470150-726
Supplier: Epredia
Supplier #: 21016-06-001
Jump to:
Product Details & Documents
Protect skin from microbes and dryness with this antimicrobial strength soap that is extremely mild on hands.
- Replenishes natural oils
- 16 oz pump bottle
- Effective against gram-positive and gram-negative bacteria
The unique formulation helps replenish the skin’s natural oils that are rinsed away by frequent washing. Soap contains 1% chloroxylenol (PCMX), a fast-acting broad spectrum antimicrobial ingredient, effective against gram-negative and gram-positive bacteria, as well as yeast. 16oz pump bottle.
Specifications
- Description:VWR® softcide® extra-mild hand soap