Orders LinkContactUsLinkComponent
VWR® Measuring Pipettes
VWR® Measuring Pipettes
VWR® Measuring Pipettes
Supplier:  VWR International
Certificates
VWR® Measuring Pipettes
VWR® Measuring Pipettes
4 Options Available Below
Product Details & Documents
These high-quality borosilicate, Mohr-type pipettes meet ASTM standards.
  • Color-coded by size
  • Calibrated 'to deliver'
  • Measurements are accurate to ±0.01 ml
The pipettes feature colored graduations in 0.1 increments that are enameled into the glass to prevent fading.
Documents
Product Family Options
Product Information
  • Capacity
    Subdivisions
    Availability
    Price
  • 470149-282
    VWR® Measuring Pipette
    VWR® Measuring Pipette
    1 ml
    0.1 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 6
    Paquete de 6 - $401.56
    $401.56
    Log in for availability
    Case de 2 (6UN)
    Case de 2 (6UN) - $437.70
    $437.70
    Paquete de 6
       $401.56
    Case de 2 (6UN)
       $437.70
  • 470149-280
    VWR® Measuring Pipette
    VWR® Measuring Pipette
    5 ml
    0.1 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 6
    Paquete de 6 - $385.84
    $385.84
    Log in for availability
    Case de 2 (6UN)
    Case de 2 (6UN) - $420.56
    $420.56
    Paquete de 6
       $385.84
    Case de 2 (6UN)
       $420.56
  • 470149-278
    VWR® Measuring Pipette
    VWR® Measuring Pipette
    10 ml
    0.1 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 6
    Paquete de 6 - $385.84
    $385.84
    Log in for availability
    Case de 2 (6UN)
    Case de 2 (6UN) - $420.56
    $420.56
    Paquete de 6
       $385.84
    Case de 2 (6UN)
       $420.56
  • 470149-276
    VWR® Measuring Pipette
    VWR® Measuring Pipette
    25 ml
    0.1 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 6
    Paquete de 6 - $686.05
    $686.05
    Log in for availability
    Case de 2 (6UN)
    Case de 2 (6UN) - $747.79
    $747.79
    Paquete de 6
       $686.05
    Case de 2 (6UN)
       $747.79