VWR® Liquid Nitrogen Dewars
Flexible Option for Safe Handling of Chemicals.
- Durable
- Available in a range of volumes
Safely store and dispense small and large quantities of liquid nitrogen. High strength neck tube reduces liquid nitrogen loss. Optional pour spout is available to increase control and handling. A sturdy five-wheel rollerbase with locking casters for the 30L VWR® Liquid Nitrogen Dewar is available separately.
- Catalog No:
- 470128-892
- 470201-128
- Capacidad:
- 5 L
- 10 L
Frequently Bought Together