VWR® Linear Spring Scales
Supplier: VWR International
Jump to:
Acerca de este artículo
Designed for Heavy Duty Student Use
- Feature easy to read markings
- Durable for years of regular student use
- Length: 14 cm
- Each scale is easily zeroed
Each has a plated steel suspension ring at the top, and a hook at the bottom.
5 Options Available Below
Product Details & Documents
Designed for Heavy Duty Student Use
- Feature easy to read markings
- Durable for years of regular student use
- Length: 14 cm
- Each scale is easily zeroed
Each has a plated steel suspension ring at the top, and a hook at the bottom.
Product Family Options
Información del producto
- Weighing CapacityAvailabilityPrice
- 250 g x 5 g (2.5 N x 0.05 N)Discontinued-
- 500 g x 10 g (5 N x 0.1 N)Discontinued-
- 1000 g x 20 g (10 N x 0.2 N)Discontinued-
- 2000 g x 40 g (20 N x 0.4 N)Each (1UN)$3.35Case de 100 (1UN)$335.00
- 5000 g x 100 g (50 N x 5 N)Discontinued-
- 250 g x 5 g (2.5 N x 0.05 N)Each (1UN)$3.35Case de 100 (1UN)$335.00
- 500 g x 10 g (5 N x 0.1 N)Each (1UN)$110.70Case de 100 (1UN)$120.66
- 1000 g x 20 g (10 N x 0.2 N)Each (1UN)$111.62Case de 100 (1UN)$121.66
- 5000 g x 100 g (50 N x 5 N)Each (1UN)$3.35Case de 100 (1UN)$335.00