VWR® Disposable Spatulas
Standard disposable spatula, double-ended spoon and scoop, PP, Opaque, 21 cm (8¼"), Non sterile
Supplier: VWR International
Jump to:
Product Details & Documents
Look for the V at the neck.™ Economical, disposable tools for transferring, collecting, and handling chemical and biological materials.
- Economical, polypropylene, single-use laboratory spatulas
- Ideal for transferring, manipulating, and mixing materials
- Versatile double-ended design
- Help minimize cross contamination
- Standard (double-ended spoon/scraper and scoop) fits 1.5 ml and 15 ml tubes
- SGS tested for compliance with US and EU regulations for food-contact applications
- Resistant to dilute acids and bases
- Autoclavable, 15 psi for 15 min at 121 °C
Certifications: Manufactured In compliance with US and EU food contact standards. For research only, not a medical product.
Packaging: Nonsterile spatulas are packed in a folded closed poly bag in a paperboard carton.
The V design at the neck of the disposable laboratory spatula is a trademark of LevGo, Inc.
Caution: Not for medical use.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Sterility:Non sterile
- Color:Opaque
- Description:Standard disposable spatula, double-ended spoon and scoop
- Length:21 cm (8¼")
- Material:PP