VWR® Cylinders with Plastic Base
Supplier: VWR International
Acerca de este artículo
Our high-quality borosilicate cylinders meet ASTM standards and are a great value.
- Double graduated metric scale
- Pour-out
- Detachable plastic base
Featuring uniform white enamel, they can be heated and used with acids, salt solutions, and organic solvents.
4 Options Available Below
Product Details & Documents
Our high-quality borosilicate cylinders meet ASTM standards and are a great value.
- Double graduated metric scale
- Pour-out
- Detachable plastic base
Featuring uniform white enamel, they can be heated and used with acids, salt solutions, and organic solvents.
Documents
Certificates
Buscar por número de loteSpecifications for Product Family
Especificaciones
- Catalog No:
- 470191-190
- 470191-206
- 470191-192
- 470191-194
- Form:
- Tall
- Tall
- Tall
- Tall
Especificaciones
Product Family Options
Información del producto
- CapacityAvailabilityPrice
- 10 mlEach (1UN)$48.90Case de 12 (1UN)$53.30Especificaciones:
More
Specifications
Cat. No.470191-190FormTall - 25 mlEach (1UN)$58.58Case de 12 (1UN)$63.85Especificaciones:
More
Specifications
Cat. No.470191-206FormTall - 50 mlEach (1UN)$68.80Case de 12 (1UN)$74.99Especificaciones:
More
Specifications
Cat. No.470191-192FormTall - 100 mlEach (1UN)$113.40Case de 12 (1UN)$123.61Especificaciones:
More
Specifications
Cat. No.470191-194FormTall