Volvox Slide
Volvox Slide, Section, Quadruple
Catalog # 470180-992
Supplier: Avantor
Jump to:
Product Details & Documents
Freshwater green algae
- Identifying features clearly distinguishable
- Rigorous quality control standards
Freshater chlorophytes. Asexual and sexual reproductive stages.
Especificaciones
- Description:Volvox Slide
- Stain:Quadruple
- Orientation:Section