Orders LinkContactUsLinkComponent
PYREX® Volumetric Flasks with [ST] Stopper
PYREX® Volumetric Flasks with [ST] Stopper
PYREX® Volumetric Flasks with [ST] Stopper
PYREX® Volumetric Flasks with [ST] Stopper
Made of PYREX® borosilicate glass.
  • Graduation line is sharp and permanent
  • Large white block letters make inscriptions easy to read
  • Supplied with interchangeable ground glass stoppers
Flasks are designed for those that do not require the precision accuracy of our Class A capacity flasks. Capacity tolerances are twice those specified for Class A volumetric ware.
Product Family Options
  • Capacity
    Description
    ST Stopper No.
    Tolerance
  • 100 ml
    Pyrex® Volumetric Flasks with Glass Stopper, 100 ml
    13
    ±0.16 ml
    Each (1UN)
    Each (1UN) - $1,320.00
    $1,320.00
    Case de 12 (1UN)
    Case de 12 (1UN) - $1,403.62
    $1,403.62
    Each (1UN)
       $1,320.00
    Case de 12 (1UN)
       $1,403.62
     : 5641-100
  • 500 ml
    Pyrex® Volumetric Flasks with Glass Stopper, 500 ml
    19
    ±0.40 ml
    Each (1UN)
    Each (1UN) - $2,020.00
    $2,020.00
    Case de 12 (1UN)
    Case de 12 (1UN) - $2,157.59
    $2,157.59
    Each (1UN)
       $2,020.00
    Case de 12 (1UN)
       $2,157.59
     : 5641-500