Orders LinkContactUsLinkComponent
Vinyl Disposable Gloves
Vinyl Disposable Gloves
Supplier:   3G PACKAGING, INC.
Vinyl Disposable Gloves
Supplier:  3G PACKAGING, INC.
Certificates
Vinyl Disposable Gloves
4 Options Available Below
Product Details & Documents
These strong, thin, form fitting gloves are designed for a skin-tight fit on either hand.
  • Ambidextrous
  • Non sterile
  • Latex-Free
  • Multiple Sizes
  • Pre-Powdered Option
They are designed to have the same fit and protection as latex gloves without the irritation that affects some people.
They are comfortable with a sensitive response and available in your choice of sizes. A ore-powered option is available for easy on and off.
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 470122-432
  • 470122-434
  • 470122-436
  • 470153-640
  • 470018-304
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powdered
  • Powdered
Especificaciones
Product Family Options
Información del producto
  • Glove size
    Glove Color
    Length
    Packaging
    Availability
    Price
  • 470122-432
    Powder-free vinyl
    Powder-free vinyl
    S
    Translucent
    22.8 cm (9")
    100 gloves/box
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $289.16
    $289.16
    Log in Para disponibilidad
    Case de 10 (100UN)
    Case de 10 (100UN) - $315.18
    $315.18
    Paquete de 100
       $289.16
    Case de 10 (100UN)
       $315.18
    Supplier Number: PV600SM
    Especificaciones:
    More
    Specifications
    Cat. No.
    470122-432
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Powdered / powder-free
    Powder-free
  • 470122-434
    Powder-free vinyl
    Powder-free vinyl
    M
    Translucent
    22.8 cm (9")
    100 gloves/box
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $289.16
    $289.16
    Log in Para disponibilidad
    Case de 10 (100UN)
    Case de 10 (100UN) - $315.18
    $315.18
    Paquete de 100
       $289.16
    Case de 10 (100UN)
       $315.18
    Supplier Number: PV600MD
    Especificaciones:
    More
    Specifications
    Cat. No.
    470122-434
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Powdered / powder-free
    Powder-free
  • 470122-436
    Powder-free vinyl
    Powder-free vinyl
    L
    Translucent
    22.8 cm (9")
    100 gloves/box
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $289.16
    $289.16
    Log in Para disponibilidad
    Case de 10 (100UN)
    Case de 10 (100UN) - $315.18
    $315.18
    Paquete de 100
       $289.16
    Case de 10 (100UN)
       $315.18
    Supplier Number: PV600LG
    Especificaciones:
    More
    Specifications
    Cat. No.
    470122-436
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Powdered / powder-free
    Powder-free
  • 470153-640
    Powdered vinyl
    Powdered vinyl
    S
    Translucent
    22.8 cm (9")
    100 gloves/box
    Discontinued
    Supplier Number: GVDR-SM-1C
    Especificaciones:
    More
    Specifications
    Cat. No.
    470153-640
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Powdered / powder-free
    Powdered
  • 470018-304
    Powdered vinyl
    Powdered vinyl
    M
    Translucent
    22.8 cm (9")
    100 gloves/box
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Caja de 100
    Caja de 100 - $339.14
    $339.14
    Log in Para disponibilidad
    Case de 10 (100UN)
    Case de 10 (100UN) - $3,391.43
    $3,391.43
    Caja de 100
       $339.14
    Case de 10 (100UN)
       $3,391.43
    Supplier Number: GVDR-MED
    Especificaciones:
    More
    Specifications
    Cat. No.
    470018-304
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Powdered / powder-free
    Powdered
Accessories
Información del producto
  • Catalog #
    Glove size
    Glove Color
    Length
    Availability
    Price
  • 470150-920
    XL
    Translucent
    22.8 cm (9")
    Log in Para disponibilidad
    Paquete de 100 - $522.33