Orders LinkContactUsLinkComponent
Vinyl Lab Apron
Vinyl Lab Apron
Vinyl Lab Apron
Certificates
Vinyl Lab Apron
3 Options Available Below
Product Details & Documents
Tough vinyl lab aprons are resistant to lab reagents, acids, alkalis, oils, and grease.
  • Halter Design
  • Aprons Do Not Restrict Movement
  • Two Sizes Available
  • Resistant to Acids, Alkalis, Oils, and More
  • Reinforced Strain Points
  • Latex free
The halter design keeps arms free for unrestricted movement in the lab.
The aprons are reinforced at strain points.
Product Family Options
Información del producto
  • Color
    W×L
    Availability
    Price
  • 470148-718
    Adult Apron
    Adult Apron
    Clear
    35×45"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $68.26
    $68.26
    Log in Para disponibilidad
    Case de 50 (1UN)
    Case de 50 (1UN) - $74.40
    $74.40
    Each (1UN)
       $68.26
    Case de 50 (1UN)
       $74.40
    Supplier Number: VNAP-AD
  • 470148-664
    Child Apron
    Child Apron
    Clear
    27×36"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $48.37
    $48.37
    Log in Para disponibilidad
    Case de 50 (1UN)
    Case de 50 (1UN) - $52.73
    $52.73
    Each (1UN)
       $48.37
    Case de 50 (1UN)
       $52.73
    Supplier Number: VNAP-CH
  • 470228-492
    X-Small Apron
    X-Small Apron
    Clear
    15×22"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $136.45
    $136.45
    Each (1UN)
       $136.45
    Supplier Number: VNAPXS