Vagina Slide
Vagina Slide, Longitudinal section, Hematoxylin and Eosin
Catalog # 470183-116
Supplier: Avantor
Jump to:
Product Details & Documents
Female reproductive system
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human. General structure of walls; stratified squamous epithelium with thick smooth muscle layer.
Especificaciones
- Description:Vagina Slide
- Stain:Hematoxylin and Eosin
- Orientation:Longitudinal section