Orders LinkContactUsLinkComponent
Vacuum Chamber
Vacuum Chamber, H x OD, in.: 6-5/8 x 9-3/8
  470211-850
 :  5305-0609
Vacuum Chamber
  470211-850
Vacuum Chamber
Vacuum Chamber
Vacuum Chamber, H x OD, in.: 6-5/8 x 9-3/8
$6,920.00
Each (1UN) Case de 2 (1UN)
Nalgene Complete polyetherimide jar and vacuum plate chamber provides strength and good chemical resistance.
Neoprene gasket provides secure seal. Slight amber cast of jar allows complete visibility of interior.
Plate has a center hole for exhaust with tube leading to an external hose connector which accepts 1/4" ID tubing.
For use at room temperature and at full vacuum.
  • Description:
    Vacuum Chamber, H x OD, in.: 6-5/8 x 9-3/8