Utility Shears
Utility Shears
Catalog # 470007-988
Supplier: HEALTH SMART SE
Número de proveedor #: 27-755-020
Jump to:
Product Details & Documents
Cut everything from paper to 24-gauge stainless steel with these shears.
- Lightweight, 420 stainless steel blades (Rockwell hardness of 500)
- Autoclavable polypropylene handles
Length: 9"
Especificaciones
- Description:Utility Shears