Uterus, Menstrual Phase Slide
Uterus, Menstrual Phase Slide, Section, Hematoxylin and Eosin
Catalog # 470183-112
Supplier: Avantor
Jump to:
Product Details & Documents
Female reproductive system
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human. Demonstrates ischaemia and degenerating endometrium.
Especificaciones
- Description:Uterus, Menstrual Phase Slide
- Stain:Hematoxylin and Eosin
- Orientation:Section