ContactUsLinkComponent
Usage of Restriction Enzymes in DNA Fingerprinting Analysis
Usage of Restriction Enzymes in DNA Fingerprinting Analysis
Catálogo # 470126-554
Proveedor:  EDVOTEK, INC.
undefined
Usage of Restriction Enzymes in DNA Fingerprinting Analysis
Catálogo # 470126-554
Proveedor:  EDVOTEK, INC.
Número de proveedor:  225
Especificaciones
  • Descripción:
    Usage of Restriction Enzymes in DNA Fingerprinting Analysis
  • No. of Lab Groups:
    6
  • Referencia:
    470126-554
  • Nº de proveedor:
    225
Especificaciones
Acerca de este artículo
Compare DNA Banding Patterns
  • Time required: ~90 min.
Teach your students about restriction enzyme digests in the context of forensic science! Your students will cut DNA with restriction enzymes and then compare the banding pattern of the crime scene DNA versus that of two suspects using agarose gel electrophoresis.
Kit includes: instructions, “crime scene” Ready-to-Load™ DNA samples, Standard DNA Marker, Dryzymes® - Eco RI and Hind III, various solutions & buffers, plasmid DNA, enzyme grade water, agarose powder, InstaStain® Blue & FlashBlue™ Stain. Kit ships directly from the vendor.
Required, but not included: electrophoresis apparatus, power supply, automatic pipette with tips, water bath, balance, microwave or hot plate, visualization (white light), misc. labware, pipette pumps or bulbs, metric rulers, floating racks, photo documentation system (optional), distilled or deionized water, ice.
Información de pedido: The kit will ship separately from the perishable component. Kits generally ship via UPS ground. Perishable components will ship via overnight or second day delivery.
Entrega: This item contains a coupon for live or perishable materials. Please redeem the coupon from the vendor and provide the date you wish the perishable components to arrive.
Embalaje: Some components require refrigerator and freezer storage on arrival.