VWR® Standard Sieve Set
Wentworth scale brass sieve set
Catalog # 470145-666
Supplier: Avantor
Jump to:
Product Details & Documents
The most durable sieve set on the market.
- Made of rust-free polished and lacuqered brass
- 8" diameter conforms to ASTM E-11 standards
- 2" deep
- Openings are indicated in milimeters and inches
These sieves stack together to accommodate any type of sample.
Ordering information: Each kit contains a set of 7 brass sieves as well as bottom and cover. Sieve sizes are: 5, 10, 18, 35, 60, 120, and 230.
Especificaciones
- Description:Wentworth scale brass sieve set