Orders LinkContactUsLinkComponent
Urinary Test Strips
Urinary Test Strips
Urinary Blood Test Strips, Vial of 100
Catalog # 470019-032
Supplier:   LW SCIENTIFIC SE
Número de proveedor #:  URS-01PR-BL77
Urinary Test Strips
Catalog # 470019-032
Supplier:  LW SCIENTIFIC SE
CAS Number:  
Urinary Test Strips
Urinary Test Strips
Urinary Blood Test Strips, Vial of 100
CAS Number:  
Price
$279.82
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Includes Color Key for Easy Reading of Results
Simply place a drop of urine on the strip’s indicator spot and wait one minute for the color change. These strips are clinically tested for accuracy and have a shelf life of two years if kept in the resealable dry containers. They can also be used with WARD’S Artificial Urine. Vials of 100.
Especificaciones
Especificaciones
  • Description:
    Urinary Blood Test Strips, Vial of 100
Especificaciones