Urinary Bladder, Distended Slide
Urinary Bladder, Distended Slide, Section, Hematoxylin and Eosin
Catalog # 470182-932
Supplier: Avantor
Jump to:
Product Details & Documents
Female reproductive system
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Demonstrates transitional epithelium, lamina propria, and muscularis of a distented bladder.
Specifications
- Description:Urinary Bladder, Distended Slide
- Stain:Hematoxylin and Eosin
- Orientation:Section