Unknown Resistance Board
Unknown Resistance Board
Catalog # 470016-288
Supplier: VWR International
Jump to:
Product Details & Documents
Use this set of nine unknown resistances to work with your Wheatstone bridges.
- Connect in series or parallel
Each resistor has a unique resistance, but all are 1/4 watt metal film with 1% accuracy. Resistances start with 1.21 KΩ to protect against overloads and range up to 90.9 KΩ at no particular interval. They are mounted visibly underneath a printed circuit board with connections through binding posts on top; the resistors are numbered 1 to 9.
Specifications
- Description:Unknown Resistance Board