Orders LinkContactUsLinkComponent
Universal Specific Gravity Kit
Universal Specific Gravity Kit
Catalog # 470018-684
Supplier:   MINERALAB, LLC
 :  SGK-C
Universal Specific Gravity Kit
Catalog # 470018-684
Supplier:  MINERALAB, LLC
Universal Specific Gravity Kit
Universal Specific Gravity Kit
Universal Specific Gravity Kit
Price
$4,157.46
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Measure the Specific Gravity of Gemstones and Minerals with Ease
Combine any electronic scale with a minimum capacity of 15 g (available separately), a tare function, and a readability of at least 0.001 g with this kit to quickly test specific gravity of rocks and minerals. Simply weigh the sample in air on the integrated weighing platform of the scale, then place the sample in the underwater weighing pan included with the kit and weigh it underwater. The kit also includes a sample holder support, a beaker and beaker support, test weight, tweezers, and instructions.
Connect to Quality and Precision in this Activity with Ohaus®
Ohaus balances, scales, and equipment meet the highest quality standards and are proven to be reliable in your classroom for years of use. Use the Ohaus Scout™ Pro Electronic Balance, 120g x 0.001g, item number 156279, with this activity to measure the specific gravity of rocks and minerals using the balance's density/specific gravity application.
Especificaciones
  • Description:
    Universal Specific Gravity Kit