Ungraduated Thermometers
Ungraduated Thermometers
470104-552
: BR/2496 PU
Make your own thermometers.
Precision made 12.5" glass tubes with a range of 0 to 245 °F. Filled with a special non-toxic oil that will not fade or separate. Pre-tested for accuracy. Package of six.
- Description:Ungraduated Thermometers