Unbreakable Mirror
Plastic mirror, 75×125 mm
Catalog # 470092-754
Supplier: CURBELL PLASTICS
Supplier #: 3IN X 5IN
Jump to:
Product Details & Documents
Excellent quality, plexiglass mirror that is unbreakable.
- Ideal for student use in any experiment
- 3 x 5 inches
- Clips are sold separately, item 470008-058
Specifications
- Description:Plastic mirror, 75×125 mm