U-Form Electromagnet
U-Form Electromagnet
Catalog # 470120-696
Supplier: VWR International
Jump to:
Product Details & Documents
Quality electromagnet.
- Wound with insulated copper wire
- Provided with barrel connections
This unique electromagnet consists of an iron U-shaped core with armature and a carrying hook. The electromagnet is designed for use on 4–6 volts DC.
Especificaciones
- Description:U-Form Electromagnet