To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Descripción:Two-Piece, Polycarbonate Lens Safety Shield
- Referencia:470003-112
- Nº de proveedor:1216
This shield is a must where chemical splashes can occur.
- Viewing area is ⅜" thick
- Base size: 12×14"
A dual coated lens provides scratch and chemical resistance. Frosted edges alert students to protective boundary. Slanted viewing surface provides unobstructed visibility. Top edges extend over the rear of the chemically-resistant, polypropylene base. Supplies with chemical splash goggles for added protection.