Twistit® Stopper Assortment
Twistit® Stopper Assortment
Catalog # 470151-194
Supplier: PLASTICOID CO INC THE SE
Número de proveedor #: ZM299ASST-B
Jump to:
Product Details & Documents
Solid, One-Hole, Two-Hole, and Three-Hole Rubber Stoppers in One Unit.
- Easily converts from solid to up to 3-hole stopper
- Available in sizes 3 to 10
- 1 lb. bag containing 33 stoppers
Consists of nine size 3 stoppers, eight size 4, six size 5, four size 6, two size 7, two size 8, one size 9 and one size 10 stopper. Sizes 3 and 4 stoppers have 2 holes maximum.
Certifications: CAUTION: Item contains latex, which is known to cause allergic reactions in some people. Use with caution.
Especificaciones
- Description:Twistit® Stopper Assortment