Tripod Stands
Tripod Stands, Inside Diameter: 4 1/2", 6"
Catalog # 470123-750
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: TRS6X9
Jump to:
Product Details & Documents
A must have for every lab.
- Cast iron, enameled finish
- Removable zinc plated legs
Good quality, enameled cast iron tripods for supporting small objects over a burner. The legs detach for easy storage.
Especificaciones
- Ext.Ø:6"
- Description:Tripod Stands, Inside Diameter: 4 1/2"