Tripod Ring Stands
Tripod Ring Stand, 3 1/2" dia., One Ring
Catalog # 470019-528
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: TRS4X9
Jump to:
Product Details & Documents
Support stand for holding labware over a burner.
- Cast iron ring
- Detachable legs
- Zinc plated legs
Hold beakers and flasks at a safe distance from the heat source such as a bunsen burner using these tripods. Each comes standard with one ring and detachable legs. Size: 9"H.
Especificaciones
- Description:Tripod Ring Stand, 3 1/2" dia., One Ring