Orders LinkContactUsLinkComponent
Triangular Supports with Rods
Triangular Supports with Rods
Certificates
Triangular Supports with Rods
Triangular Supports with Rods
3 Options Available Below
Product Details & Documents
Heavy cast-iron triangular bases.
  • Low flat legs on triangular supports provide excellent stability
Black enamel finish is extremely resistant to wear.
 : Includes a removable, zinc-plated steel rod.
Product Family Options
Información del producto
  • Base Dimensions
    Rod Diameter
    Rod Length
    Availability
    Price
  • 4"
    ⅜"
    20"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $389.63
    $389.63
    Each (1UN)
       $389.63
    Supplier Number: TSR420
  • 6"
    ½"
    36"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $608.70
    $608.70
    Each (1UN)
       $608.70
    Supplier Number: TSR636
  • 5"
    ½"
    24"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $538.87
    $538.87
    Each (1UN)
       $538.87
    Supplier Number: TSR524