Orders LinkContactUsLinkComponent
Triangular Files
Triangular Files
Certificates
Triangular Files
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Useful for metal filing or glass cutting.
  • Wooden handle for easy gripping
  • Hardened triple sides
Triangular file has a tapered shape, and is mounted on a hardwood handle for easy gripping. For general laboratory use, including to score glass before cutting.
Product Family Options
Product Information
  • Length
    Overall Length
    Availability
    Price
  • 470005-474
    Triangular file
    Triangular file
    File: 4"
    7"
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $153.52
    $153.52
    Each (1UN)
       $153.52
    Supplier Number: TRF004
  • 470122-172
    Triangular slim taper file with wooden handle
    Triangular slim taper file with wooden handle
    File: 7"
    9"
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $160.97
    $160.97
    Each (1UN)
       $160.97
    Supplier Number: TRF005
  • 470104-232
    Triangular file
    Triangular file
    File: 9"
    11"
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $187.09
    $187.09
    Each (1UN)
       $187.09
    Supplier Number: TRF006
Accessories
Product Information
  • Catalog #
    Length
    Overall Length
    Availability
    Price
  • Triangular file
    Triangular file
    470005-474
    File: 4"
    7"
    Log in for availability
    Each (1UN) - $153.52
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.