Tri Pour Beakers
Supplier: DYN-A-MED PRODUCTS
Jump to:
Acerca de este artículo
These unbreakable polypropylene beakers feature a tri-cornered lip for dripless pouring.
Accurate graduations to ±5
- Tri-cornered lip for dripless pouring
- Unbreakable
- Resistant to common acids, solvents, and alkalis
- Autoclavable
They are unaffected by commonly used acids, alkalis, and solvents and are autoclavable.
Graduations are accurate to ±5.
1 Options Available Below
Product Details & Documents
These unbreakable polypropylene beakers feature a tri-cornered lip for dripless pouring.
Accurate graduations to ±5
- Tri-cornered lip for dripless pouring
- Unbreakable
- Resistant to common acids, solvents, and alkalis
- Autoclavable
They are unaffected by commonly used acids, alkalis, and solvents and are autoclavable.
Graduations are accurate to ±5.
Product Family Options
Información del producto
- CapacityAvailabilityPrice
- 470174-588Tri-pour beakersTri-pour beakers50 ml (1.7 oz.)Discontinued-Supplier Number: 80092
- 470174-586Tri-pour beakersTri-pour beakers100 ml (3.4 oz.)Discontinued-Supplier Number: 80093
- 470174-582Tri-pour beakersTri-pour beakers400 ml (14 oz.)Discontinued-Supplier Number: 80096
- 470174-590Tri-Pour Beakers, Set of 6 different sizesTri-Pour Beakers, Set of 6 different sizes50 ml, 100 ml, 250 ml, 400 ml, 800 ml, 1000 mlPaquete de 6$287.54Case de 5 (6UN)$313.42Supplier Number: 80091