Orders LinkContactUsLinkComponent
Lab-Aids® Tree Ring Dating Kit
Lab-Aids® Tree Ring Dating Kit
Tree Ring Dating Kit, 12
Catalog # 470112-528
Número de proveedor #:  52
Lab-Aids® Tree Ring Dating Kit
Catalog # 470112-528
Lab-Aids® Tree Ring Dating Kit
Lab-Aids® Tree Ring Dating Kit
Tree Ring Dating Kit, 12
Price
$2,544.58
Log in Para disponibilidad
Kit de 1
Product Details & Documents
Explore dendrochronology.
  • Determine the age of the trees
  • Compare wet and dry years
  • Establish the year lumber was cut and when it started growing
  • Understand tree-ring dating
Students develop a basic knowledge of dendrochronology using the specially designed simulated tree cores in this engaging kit. Your students will learn the important effects of climate on living things as they unravel secrets about the age and history of trees. They will also be asked to estimate the climate of a particular year in the distant past by carefully studying a tree's annual rings. Students can also infer similar information by using various wooden objects. This Lab-Aid uses highly motivating, easily performed activities. It is recommended for grades five through college. Materials in this kit, designed for 30 students, are reusable. Following a carefully designed lab activity using two different tree core samples.
Delivery information: See datasheet for contents information.
Documents
Especificaciones
  • Description:
    Tree Ring Dating Kit
  • No. of Lab Groups:
    12