Orders LinkContactUsLinkComponent
Trebuchet
Trebuchet
Trebuchet
Catalog # 470007-314
Supplier:   EN VISION LABS SE
Número de proveedor #:  10200
Trebuchet
Catalog # 470007-314
Supplier:  EN VISION LABS SE
Trebuchet
Trebuchet
Trebuchet
Price
$429.96
Log in Para disponibilidad
Each (1UN) Case de 25 (1UN)
Product Details & Documents
Trebuchet: the greatest weapon from the ancient world
  • Available as an individual kit or classroom package
  • This desktop Trebuchet kit provides a fun and exciting environment for studying energy conversion, levers, trajectories, ratios, rotational motion, inertia, forces and more!
Powered by gravitational potential energy, this easy-to-assemble kit provides hands-on, practical building experience while fueling the imagination with history and science. Investigate how projectile range is altered by changing such variables as weight, weight position, sling length, pivot point and projectile mass. Each kit is supplied with assembly instructions and Internet resource materials that provide investigations into the theory of operation, mathematical modeling, history and more!
Especificaciones
  • Description:
    Trebuchet