Tray/Tote Box
Tray/Tote Box
Catalog # 470173-908
Supplier: DYNALON
Número de proveedor #: 419244
Jump to:
Product Details & Documents
Designed to nest together without sticking, this tray is molded of hefty linear polyethylene.
- It will not corrode or mildew
- Size: 21L×12W×6"D
Remains chemically resistant to alkalis, oils, acids, and detergents.
Especificaciones
- Description:Tray/Tote Box