Orders LinkContactUsLinkComponent
Compartmented Plastic Boxes, Clear, Flambeau
Compartmented Plastic Boxes, Clear, Flambeau
Compartmented Plastic Boxes, Clear, Flambeau
Certificates
Compartmented Plastic Boxes, Clear, Flambeau
Compartmented Plastic Boxes, Clear, Flambeau
3 Options Available Below
Product Details & Documents
Keep small specimens or other items organized and visible in these clear polystyrene boxes with hinged lids.
  • Transparent
  • Butadiene copolymer material
Product Family Options
Información del producto
  • No. of Compartments
    Dimensions
    Availability
    Price
  • 18
    15.7 cm (6³/₁₆")×26.7 cm (10¹/₂")×4 cm (1⁹/₁₆") (W×L×H)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $271.41
    $271.41
    Log in Para disponibilidad
    Case de 24 (1UN)
    Case de 24 (1UN) - $295.84
    $295.84
    Each (1UN)
       $271.41
    Case de 24 (1UN)
       $295.84
    Supplier Number: K618
  • 18
    15.7 cm (6³/₁₆")×26.7 cm (10¹/₂")×4 cm (1⁹/₁₆") (W×L×H)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $471.88
    $471.88
    Log in Para disponibilidad
    Case de 24 (1UN)
    Case de 24 (1UN) - $514.35
    $514.35
    Each (1UN)
       $471.88
    Case de 24 (1UN)
       $514.35
    Supplier Number: K618
  • 24
    21.6 cm (8¹/₂")×32.4 cm (12³/₄")×5.4 cm (2¹/₈") (W×L×H)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $782.53
    $782.53
    Log in Para disponibilidad
    Case de 12 (1UN)
    Case de 12 (1UN) - $852.96
    $852.96
    Each (1UN)
       $782.53
    Case de 12 (1UN)
       $852.96
    Supplier Number: A824
  • 24
    21.6 cm (8¹/₂")×32.4 cm (12³/₄")×5.4 cm (2¹/₈") (W×L×H)
    Discontinued
    Supplier Number: K824