Top Shelf Human Anatomy And Physiology
470023-098
- Descripción:Top Shelf Human Anatomy And Physiology
- Longitud:21 cm
- Referencia:470023-098
- No. of Pages:165
- Anchura:27 cm
- Nº de proveedor:082515510X
Dawn Hudson
Guide students through a series of hands-on experiments aligned to national standards and designed for the high school anatomy and physiology curriculum. Inquiry based labs allow students to apply knowledge from other areas of study and develop critical thinking skills. Various methodologies are employed in each activity outlined and in-depth background information is provided for each subject area. Beginning with a basic introduction to anatomy, this activity book travels through the cells and tissues, the integumentary, skeletal, nervous, endocrine, digestive, cardiovascular, immune, reproductive, respiratory, and urinary systems. A glossary and appendices with additional resources are included.