Tirrill Burners
Supplier: VWR International
Jump to:
Acerca de este artículo
Precise Flame Control.
- Gas Inlet: 3/8" O.D.
- Mixing Tube: 1/2" O.D.
- Height: 51/4"
- LP gas burner, compatible with propane
These burners allow you to adjust both the gas and air supply for precise flame control. The aluminum mixing tube features a flame stabilizer, as well as both needle valve and air ports. It has a nickel-plated zinc base.
2 Options Available Below
Product Details & Documents
Precise Flame Control.
- Gas Inlet: 3/8" O.D.
- Mixing Tube: 1/2" O.D.
- Height: 51/4"
- LP gas burner, compatible with propane
These burners allow you to adjust both the gas and air supply for precise flame control. The aluminum mixing tube features a flame stabilizer, as well as both needle valve and air ports. It has a nickel-plated zinc base.
Product Family Options
Información del producto
- BTU OutputAvailabilityPrice
- 470148-642Tirrill burnerTirrill burner800 - 1200 BTU/cu. ft.Each (1UN)$807.43Case de 10 (1UN)$880.10
- 470148-694Tirrill burnerTirrill burner2000 - 3200 BTU/cu. ft.Each (1UN)$800.80Case de 10 (1UN)$872.87