Thymus, Mammal Slide
Thymus, Mammal, Section, Hematoxylin and Eosin
Catalog # 470182-782
Supplier: Avantor
Jump to:
Product Details & Documents
Immune System Organ
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Gland in stage of involution. Important in the development of T cells.
Especificaciones
- Description:Thymus, Mammal
- Stain:Hematoxylin and Eosin
- Orientation:Section