Thymus, Mammal Slide
Thymus, Mammal Slide, Section, Hematoxylin and Eosin
Catalog # 470182-780
Supplier: Avantor
Jump to:
Product Details & Documents
Lymphoid organ
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Shows Hassall’s corpuscles and other details of the organ.
Specifications
- Description:Thymus, Mammal Slide
- Stain:Hematoxylin and Eosin
- Orientation:Section