Orders LinkContactUsLinkComponent
Thymus, Mammal Slide
Thymus, Mammal Slide
Thymus, Mammal Slide, Section, Hematoxylin and Eosin
Catalog # 470182-780
Supplier:   Avantor
Thymus, Mammal Slide
Catalog # 470182-780
Supplier:  Avantor
Thymus, Mammal Slide
Thymus, Mammal Slide
Thymus, Mammal Slide, Section, Hematoxylin and Eosin
Price
$193.60
Log in for availability
Each (1UN)
Product Details & Documents
Lymphoid organ
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Shows Hassall’s corpuscles and other details of the organ.
Specifications
  • Description:
    Thymus, Mammal Slide
  • Stain:
    Hematoxylin and Eosin
  • Orientation:
    Section