The Cartoon Guide To Genetics
470006-354
- Descripción:The Cartoon Guide To Genetics
- Longitud:19 cm
- Referencia:470006-354
- No. of Pages:224
- Anchura:23 cm
- Nº de proveedor:0062730991
Larry Gonick and Mark Wheelis
The principles of genetics are illustrated, simplified, and humor coated, to make them immediately understandable and unforgettable, making this a perfect supplement to an introductory genetics course.