The Audubon Society Field Guides
Supplier: RANDOM HOUSE INC SE
Certificates
About this item
These easy-to-use guides feature full-color photographs of living specimens as you would see them in their natural habitats, making identification of specimens a breeze.
- Full color photographs
- Detailed information
- Vinyl covers for protection in the field
In addition, the washable vinyl covers are made to withstand rough handling in the field. Each compact guide also contains a comprehensive index and glossary.
Frequently Bought Together