Testis, Mammal Slide
Testis, Mammal, Section, Hematoxylin and Eosin
Catalog # 470182-938
Supplier: Avantor
Jump to:
Product Details & Documents
Male Reproductive System
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Seminiferous tubules cut in various planes.
Especificaciones
- Description:Testis, Mammal
- Stain:Hematoxylin and Eosin
- Orientation:Section