Orders LinkContactUsLinkComponent
Test Tubes with Rim
17 ml, 15×150 mm
  470123-578
 :  TT9800-E
Test Tubes with Rim
  470123-578
Test Tubes with Rim
Test Tubes with Rim
17 ml, 15×150 mm
$1,021.79
Paquete de 72
Pre-cleaned borosilicate glass test tubes with rims. Tubes can be used for hot or cold applications, are reusable, and can be autoclaved.
  • Tubes have rims
  • Autoclavable
Made from borosilicate glass, these general-purpose tubes will withstand daily use at an affordable price. Tubes have a uniform thickness and a strong lip.
  • Capacity:
    17 ml
  • Diameter ×H:
    15×150 mm