Test Tube Clamp With Finger Grips (Stoddard Clamp)
Catalog # 470121-808
Supplier: UNITED SCIENTIFIC SUPPLIES
Specifications
- Descripción:Stoddard Clamp with Finger Grips
- Longitud:5" (12.7 cm)
- Referencia:470121-808
- Nº de proveedor:TTCL03
Specifications
About this item
Pick up test tubes easily and safely.
- Molded finger grips for easy manipulation
- Heavy gauge wire with one-piece construction
- Length: 5"
Self-closing, easily operated with one hand.