Test Tube Clamp (Stoddard Clamp)
Supplier: UNITED SCIENTIFIC SUPPLIES
Certificates
About this item
Perfect for Student Use.
- Length: 5" (127 mm)
- For use with test tubes up to 25 mm in diameter
Made of spring brass wire with curved quadrangular jaws.
Frequently Bought Together