Orders LinkContactUsLinkComponent
Test Tube Brushes
Test Tube Brushes
Supplier:   VWR International
Test Tube Brushes
Supplier:  VWR International
Certificates
Test Tube Brushes
Test Tube Brushes
6 Options Available Below
Product Details & Documents
These test tube brushes feature a radially tufted tip.
  • Heavy, Galvanized Wire
  • Radially Tufted Tip
  • Nylon or Natural Brushes Available
They also feature heavy, galvanized wire.
Nylon or natural bristles are available.
Product Family Options
Información del producto
  • Bristle Material
    Bristle Color
    Brush head Ø
    Availability
    Price
  • 470149-232
    Brush for tube diameter 13 - 18 mm
    Brush for tube diameter 13 - 18 mm
    Nylon
    White
    20 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $17.20
    $17.20
    Log in Para disponibilidad
    Paquete de 12 (1UN)
    Paquete de 12 (1UN) - $18.75
    $18.75
    Each (1UN)
       $17.20
    Paquete de 12 (1UN)
       $18.75
  • 470202-906
    For tube diameter 9.5-12.5 mm
    For tube diameter 9.5-12.5 mm
    Nylon
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $501.55
    $501.55
    Log in Para disponibilidad
    Paquete de 12 (1UN)
    Paquete de 12 (1UN) - $6,018.54
    $6,018.54
    Each (1UN)
       $501.55
    Paquete de 12 (1UN)
       $6,018.54
  • 470124-004
    For tube diameter 13 - 18 mm
    For tube diameter 13 - 18 mm
    Natural bristle
    Tan
    20 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $17.20
    $17.20
    Each (1UN)
       $17.20
  • 470202-904
    For tube diameter 9.5-12.5 mm
    For tube diameter 9.5-12.5 mm
    Natural bristle
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $45.98
    $45.98
    Log in Para disponibilidad
    Paquete de 12 (1UN)
    Paquete de 12 (1UN) - $551.76
    $551.76
    Each (1UN)
       $45.98
    Paquete de 12 (1UN)
       $551.76
  • 470149-236
    Brush for tube diameter 20 - 27 mm
    Brush for tube diameter 20 - 27 mm
    Nylon
    White
    29 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $17.20
    $17.20
    Log in Para disponibilidad
    Paquete de 12 (1UN)
    Paquete de 12 (1UN) - $18.75
    $18.75
    Each (1UN)
       $17.20
    Paquete de 12 (1UN)
       $18.75
  • 470149-234
    Brush for tube diameter 20 - 27 mm
    Brush for tube diameter 20 - 27 mm
    Nylon
    Tan
    29 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $18.29
    $18.29
    Log in Para disponibilidad
    Paquete de 12 (1UN)
    Paquete de 12 (1UN) - $19.94
    $19.94
    Each (1UN)
       $18.29
    Paquete de 12 (1UN)
       $19.94