Orders LinkContactUsLinkComponent
Test Tube Brushes
Test Tube Brushes
Test Tube Brushes
Test Tube Brushes
Test Tube Brushes
Will Not Scratch.
  • Choose between natural or nylon bristles
  • Neither the bristles or the radial end will scratch glass
Product Family Options
  • Description
    Bristle Material
    Brush Head Length
    Brush head Ø
    Total length
  • For tube diameter up to 10 mm
    Natural bristle
    76 mm
    13 mm (1/2")
    203 mm
    Each (1UN)
    Each (1UN) - $27.40
    $27.40
    Paquete de 12 (1UN)
    Paquete de 12 (1UN) - $29.87
    $29.87
    Each (1UN)
       $27.40
    Paquete de 12 (1UN)
       $29.87
     : 72001W
  • For tube diameter up to 16 mm
    Natural bristle
    89 mm
    19 mm (3/4")
    229 mm
    Each (1UN)
    Each (1UN) - $31.17
    $31.17
    Paquete de 12 (1UN)
    Paquete de 12 (1UN) - $33.98
    $33.98
    Each (1UN)
       $31.17
    Paquete de 12 (1UN)
       $33.98
     : 72002W
  • For tube diameter up to 10 mm
    Nylon
    76 mm
    13 mm (1/2")
    203 mm
    Each (1UN)
    Each (1UN) - $59.41
    $59.41
    Paquete de 144 (1UN)
    Paquete de 144 (1UN) - $64.75
    $64.75
    Each (1UN)
       $59.41
    Paquete de 144 (1UN)
       $64.75
     : E001
  • For tube diameter up to 16 mm
    Nylon
    89 mm
    19 mm (3/4")
    229 mm
    Each (1UN)
    Each (1UN) - $59.67
    $59.67
    Paquete de 12 (1UN)
    Paquete de 12 (1UN) - $65.04
    $65.04
    Case de 144 (1UN)
    Case de 144 (1UN) - $8,591.99
    $8,591.99
    Each (1UN)
       $59.67
    Paquete de 12 (1UN)
       $65.04
    Case de 144 (1UN)
       $8,591.99
     : E002
  • For tube diameter 16 - 25 mm
    Nylon
    102 mm
    25 mm (0.98")
    254 mm
    Each (1UN)
    Each (1UN) - $67.71
    $67.71
    Paquete de 12 (1UN)
    Paquete de 12 (1UN) - $73.80
    $73.80
    Caja de 144 (1UN)
    Caja de 144 (1UN) - $9,750.36
    $9,750.36
    Each (1UN)
       $67.71
    Paquete de 12 (1UN)
       $73.80
    Caja de 144 (1UN)
       $9,750.36
     : E004