Taylor Sling Psychrometer
Taylor Sling Psychrometer, Replacement Thermometer (F), Permacolor-Filled
Catalog # 470039-420
Supplier: PECO SALES INC SE
Supplier #: 89291
Jump to:
Product Details & Documents
Obtain quick and accurate wet and dry-bulb temperatures with this handy instrument.
Two thermometers, recessed in a gray metal back and attached to a folding swivel handle, provide temperature readings from -20° to 120°F in 1° increments. With the cloth wick on one of the thermometers, simply spin the unit to obtain respective temperatures. Includes spare wicks and instructions. Replacement permacolor-filled thermometers are available.
Specifications
- Description:Taylor Sling Psychrometer, Replacement Thermometer (F), Permacolor-Filled